Learn More
Medchemexpress LLC Indirubin-3'-oxime | 667463-82-3 | 99.1% | 277.28 | 100 MG
SDP
Catalog No. 5000352378
Encompass_Preferred
Indirubin-3'-oxime (IDR3O) is a synthetic derivative of indirubin, acting as a potent inhibitor of cyclin-dependent kinases (CDKs) and glycogen synthase kinase 3β (GSK3β). It directly inhibits all three isoforms of JNK (JNK1, JNK2, and JNK3). This product can enhance height growth by activating Wnt/β-catenin signaling in chondrocytes and prevents cerebellar granule neurons from apoptosis.
  • Inhibitor of cyclin-dependent kinases (CDKs)
  • Inhibits glycogen synthase kinase 3β (GSK3β)
  • Directly inhibits JNK1, JNK2, and JNK3 isoforms
  • Enhances height growth via activation of Wnt/β-catenin signaling
  • Blocks c-Jun phosphorylation induced by potassium withdrawal
  • Prevents cerebellar granule neurons from apoptosis
  • Significantly prevents 6OHDA-induced decrease of nuclear localized MEF2D expression
Catalog No. 50-003-52378 Supplier Medchemexpress LLC Supplier No. HY139254100MG
Indirubin-3'-oxime (IDR3O) is a synthetic derivative of indirubin, acting as a potent inhibitor of cyclin-dependent kinases (CDKs) and glycogen synthase kinase 3β (GSK3β). It directly inhibits all three isoforms of JNK (JNK1, JNK2, and JNK3). This product can enhance height growth by activating Wnt/β-catenin signaling in chondrocytes and prevents cerebellar granule neurons from apoptosis.
  • Inhibitor of cyclin-dependent kinases (CDKs)
  • Inhibits glycogen synthase kinase 3β (GSK3β)
  • Directly inhibits JNK1, JNK2, and JNK3 isoforms
  • Enhances height growth via activation of Wnt/β-catenin signaling
  • Blocks c-Jun phosphorylation induced by potassium withdrawal
  • Prevents cerebellar granule neurons from apoptosis
  • Significantly prevents 6OHDA-induced decrease of nuclear localized MEF2D expression