Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Indirubin-3′-oxime | 667463-82-3 | 99.1% | 277.28 | 25 MG
SDP

Catalog No. 5000350645
Encompass_Preferred

Indirubin-3′-oxime (IDR3O) is a synthetic derivative of indirubin that functions as a potent inhibitor of cyclin-dependent kinases (CDKs) and glycogen synthase kinase 3β (GSK3β). It directly inhibits all three isoforms of JNK (JNK1, JNK2, and JNK3). This compound also promotes height growth through the activation of Wnt/β-catenin signaling in chondrocytes and is intended for research use only.

  • Potent inhibitor of cyclin-dependent kinases (CDKs)
  • Potent inhibitor of glycogen synthase kinase 3β (GSK3β)
  • Directly inhibits JNK1 (IC50: 0.8 μM), JNK2 (IC50: 1.4 μM), and JNK3 (IC50: 1.0 μM)
  • Enhances height growth by activating Wnt/β-catenin signaling in chondrocytes
  • Blocks c-Jun phosphorylation in cerebellar granule neurons (CGNs)
  • Prevents 6OHDA-induced decrease of nuclear localized MEF2D expression in PC12 cells
  • Enhances tibial longitudinal growth in mice
  • For research use only

Catalog No. 50-003-50645 Supplier Medchemexpress LLC Supplier No. 25MGHY139254
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Indirubin-3′-oxime (IDR3O) is a synthetic derivative of indirubin that functions as a potent inhibitor of cyclin-dependent kinases (CDKs) and glycogen synthase kinase 3β (GSK3β). It directly inhibits all three isoforms of JNK (JNK1, JNK2, and JNK3). This compound also promotes height growth through the activation of Wnt/β-catenin signaling in chondrocytes and is intended for research use only.

  • Potent inhibitor of cyclin-dependent kinases (CDKs)
  • Potent inhibitor of glycogen synthase kinase 3β (GSK3β)
  • Directly inhibits JNK1 (IC50: 0.8 μM), JNK2 (IC50: 1.4 μM), and JNK3 (IC50: 1.0 μM)
  • Enhances height growth by activating Wnt/β-catenin signaling in chondrocytes
  • Blocks c-Jun phosphorylation in cerebellar granule neurons (CGNs)
  • Prevents 6OHDA-induced decrease of nuclear localized MEF2D expression in PC12 cells
  • Enhances tibial longitudinal growth in mice
  • For research use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.