Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indirubin-3'-oxime, Tocris Bioscience™

Catalog No. 181310
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-131-0 10 mg
18-135-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 18-131-0 Supplier R&D Systems Supplier No. 1813/10
This item is not returnable. View return policy

GSK-3β inhibitor. Also inhibits other protein kinases

Inhibitor of GSK-3β (IC50 = 190 nM) and cyclin dependent kinases (cdks; IC50 values are 0.05, 0.18 and 0.25-0.5 βM for cdk9, cdk1 and cdk2, respectively). Also reported to inhibit AMPK, LCK and SGK. Inhibits cdk5- and GSK-3β-mediated tau phosphorylation. Induces cell cycle arrest and inhibits cell proliferation.

Chemical Identifiers

CAS 160807-49-8
Molecular Formula C16H11N3O2
Molecular Weight (g/mol) 277.28
MDL Number MFCD02683594
InChI Key LDEWQRSYYHTQRA-UHFFFAOYSA-N
Synonym indirubin-3'-monoxime, indirubin-3'-oxime, indirubin-3-oxime, indirubin-3monoxime, indirubin-3'-monooxime, indirubin-3-monoxime, 3-3-hydroxyamino-1h-indol-2-yl indol-2-one, indirubin-3′-monoxime, z-1h,1'h-2,3' biindolylidene-3,2'-dione-3-oxime, 3-1,3-dihydro-3-hydroxyimino-2h-indol-2-ylidene-1,3-dihydro-2h-indol-2-one
PubChem CID 5326739
IUPAC Name 3-(hydroxyamino)-1H,2'H-[2,3'-biindol]-2'-one
SMILES ONC1=C(NC2=CC=CC=C12)C1=C2C=CC=CC2=NC1=O

Specifications

Quantity 10 mg
Formula Weight 277.28
Percent Purity >99%
Chemical Name or Material Indirubin-3'-oxime
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.