Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Indirubin-3'-oxime, Tocris Bioscience™
Description
Inhibitor of GSK-3β (IC50 = 190 nM) and cyclin dependent kinases (cdks; IC50 values are 0.05, 0.18 and 0.25-0.5 βM for cdk9, cdk1 and cdk2, respectively). Also reported to inhibit AMPK, LCK and SGK. Inhibits cdk5- and GSK-3β-mediated tau phosphorylation. Induces cell cycle arrest and inhibits cell proliferation.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 277.28 |
| Percent Purity | >99% |
| Chemical Name or Material | Indirubin-3'-oxime |
Safety and Handling
| Recommended Storage | Store at Room Temperature |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?