Learn More
Medchemexpress LLC HY-19807 5mg Medchemexpress, Indirubin-3'-monoxime CAS:160807-49-8 Purity:>98%
SDP
Catalog No. 502082187
Encompass_Preferred
Medchemexpress, HY-19807 5mg Indirubin-3'-monoxime CAS:160807-49-8 Indirubin-3'-monoxime is a potent GSK-3β inhibitor, and weakly inhibits 5-Lipoxygenase, with IC50s of 22 nM and 7.8-10 µM, respectively; Indirubin-3'-monoxime also shows inhibitory activities against CDK5/p25 and CDK1/cyclin B, with IC50s of 100 and 180 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2187 Supplier Medchemexpress LLC Supplier No. HY198075MG
Medchemexpress, HY-19807 5mg Indirubin-3'-monoxime CAS:160807-49-8 Indirubin-3'-monoxime is a potent GSK-3β inhibitor, and weakly inhibits 5-Lipoxygenase, with IC50s of 22 nM and 7.8-10 µM, respectively; Indirubin-3'-monoxime also shows inhibitory activities against CDK5/p25 and CDK1/cyclin B, with IC50s of 100 and 180 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.