Learn More
Medchemexpress LLC Indirubin Derivative E804 | 854171-35-0 | 98.5% | 365.38 | 1 ML
SDP
Catalog No. 5000388252
Encompass_Preferred
Indirubin Derivative E804 is a potent inhibitor of Insulin-like Growth Factor 1 Receptor (IGF1R), with an IC50 of 0.65 μM for IGF1R. It is a water-soluble indirubin derivative that also inhibits CDK2/CycE with an EC50 of 0.23 μM.
  • Potent inhibitor of Insulin-like Growth Factor 1 Receptor (IGF1R)
  • Water-soluble indirubin derivative
  • Inhibits CDK2/CycE
  • Suitable for research use only
Catalog No. 50-003-88252 Supplier Medchemexpress LLC Supplier No. HY1878510MM/1ML
Indirubin Derivative E804 is a potent inhibitor of Insulin-like Growth Factor 1 Receptor (IGF1R), with an IC50 of 0.65 μM for IGF1R. It is a water-soluble indirubin derivative that also inhibits CDK2/CycE with an EC50 of 0.23 μM.
  • Potent inhibitor of Insulin-like Growth Factor 1 Receptor (IGF1R)
  • Water-soluble indirubin derivative
  • Inhibits CDK2/CycE
  • Suitable for research use only