Learn More
Selleck Chemical LLC Indirubin 5mg 479-41-4 NSC 105327
SDP
Supplier:  Selleck Chemical LLC S23865MG
Encompass_Preferred
Indirubin (NSC 105327) is a potent cyclin-dependent kinases and GSK-3? inhibitor with IC50 of about 5 ?M and 0.6 ?M. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-777-1