Learn More
INDICAL INC INDISPIN PATHOGEN KIT 250
SDP
Catalog No. NC2029640
Encompass_Preferred
The IndiSpin Pathogen Kit is designed for high-quality and efficient extraction of pathogen nucleic acids, including viral RNA and DNA, and bacterial DNA. It is suitable for use with a wide variety of sample types such as whole blood, serum, plasma, other body fluids, swabs, washes, and tissue homogenates. Key features of the kit include:•A unified protocol for both DNA and RNA extraction.•Effective removal of inhibitors and contaminants for high-purity results.•Compatibility with automation on the QIAcube for low to medium throughput workflows.•A centrifugation-based protocol for straightforward implementation.The purified nucleic acids are free of proteins, nucleases, and other impurities, making them ideal for downstream applications, including real-time PCR-based pathogen detection. This versatile kit is particularly well-suited for applications in fields such as veterinary diagnostics, where sample types can vary significantly.
Catalog No. NC2029640 Supplier INDICAL INC Supplier No. SP54106
Add to Cart
Add to Cart
The IndiSpin Pathogen Kit is designed for high-quality and efficient extraction of pathogen nucleic acids, including viral RNA and DNA, and bacterial DNA. It is suitable for use with a wide variety of sample types such as whole blood, serum, plasma, other body fluids, swabs, washes, and tissue homogenates. Key features of the kit include:•A unified protocol for both DNA and RNA extraction.•Effective removal of inhibitors and contaminants for high-purity results.•Compatibility with automation on the QIAcube for low to medium throughput workflows.•A centrifugation-based protocol for straightforward implementation.The purified nucleic acids are free of proteins, nucleases, and other impurities, making them ideal for downstream applications, including real-time PCR-based pathogen detection. This versatile kit is particularly well-suited for applications in fields such as veterinary diagnostics, where sample types can vary significantly.