Learn More
Medchemexpress LLC Indisulam | 165668-41-7 | MFCD00945325 | 99.8% | 385.85 | 200 MG
SDP
Catalog No. 5000348836
Encompass_Preferred
Indisulam (E 7070) is a carbonic anhydrase inhibitor with anticancer activity. This sulfonamide agent targets the G1 phase of the cell cycle, causing a blockade in the G1/S transition. It achieves this by inhibiting the activation of both CDK2 and cyclin E. Additionally, it targets splicing by inducing RBM39 degradation via recruitment to DCAF15.
  • Carbonic anhydrase inhibitor
  • Exhibits anticancer activity
  • Sulfonamide agent
  • Targets G1 phase of the cell cycle
  • Inhibits CDK2 and cyclin E activation
  • Induces RBM39 degradation
  • Involved in splicing modulation
Catalog No. 50-003-48836 Supplier Medchemexpress LLC Supplier No. HY13650200MG
Indisulam (E 7070) is a carbonic anhydrase inhibitor with anticancer activity. This sulfonamide agent targets the G1 phase of the cell cycle, causing a blockade in the G1/S transition. It achieves this by inhibiting the activation of both CDK2 and cyclin E. Additionally, it targets splicing by inducing RBM39 degradation via recruitment to DCAF15.
  • Carbonic anhydrase inhibitor
  • Exhibits anticancer activity
  • Sulfonamide agent
  • Targets G1 phase of the cell cycle
  • Inhibits CDK2 and cyclin E activation
  • Induces RBM39 degradation
  • Involved in splicing modulation