Learn More
TARGETMOL CHEMICALS INC INDISULAM 25MG
SDP
Catalog No. 502576777
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Indisulam (E 7070) is a carbonic anhydrase inibitor and Antitumor CDK inhibitor. Indisulam targets the G1 phase of the cell cycle by depleting cyclin E. inducing p53 and p21 and inhibiting CDK2 causing a blockade in the G1/S transition. purity: 99%
Catalog No. 50-257-6777 Supplier TARGETMOL CHEMICALS INC Supplier No. T432125MG
Also available in 1 mg 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Indisulam (E 7070) is a carbonic anhydrase inibitor and Antitumor CDK inhibitor. Indisulam targets the G1 phase of the cell cycle by depleting cyclin E. inducing p53 and p21 and inhibiting CDK2 causing a blockade in the G1/S transition. purity: 99%