Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium antimonide, 99.999% (metals basis)

Catalog No. p-7049537
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1462604 2 g
AA1462609 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA1462604 Supplier Thermo Scientific Chemicals Supplier No. 01462604
Only null left
Add to Cart

CAS: 1312-41-0 | InSb | 236.578 g/mol

Indium antimonide finds use in infrared detectors, including FLIR systems, thermal imaging cameras, infrared astronomy and in infrared homing missile guidance systems, in fast transistors. It is used in thermal image detectors using photo-electromagnetic detectors or photodiodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1312-41-0
Molecular Formula InSb
Molecular Weight (g/mol) 236.578
MDL Number MFCD00016146
InChI Key WPYVAWXEWQSOGY-UHFFFAOYSA-N
PubChem CID 3468413
IUPAC Name indiganylidynestibane
SMILES [In]#[Sb]

Specifications

Melting Point 535°C
Physical Form Pieces
Quantity 2 g
UN Number UN1549
Merck Index 14,4948
Solubility Information Insoluble in water.
Formula Weight 236.57
Percent Purity 99.999%
Odor Odorless
Density 5.75 g/mL at m.p.
Assay (metals basis)
Chemical Name or Material Indium antimonide
Show More Show Less
Hazard Category H302+H332
Hazard Statement GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
EINECSNumber 215-192-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.