Learn More
TA Instruments Indium Calibration Material Not Certified, Standard Indium, Temperature Calibration, DSC, TMA, Environmental Systems, Non Certified
Catalog No. 5000580706 Shop All Waters TA Instruments Products
Encompass_Preferred
Waters TA Instruments Indium Calibration Material Not Certified is standard indium material used for temperature calibration of DSC, TMA, and other environmental systems where certified reference is not required.
Catalog No. 50-005-80706 Supplier TA Instruments Supplier No. 900902.901
Waters TA Instruments Indium Calibration Material Not Certified is standard indium material used for temperature calibration of DSC, TMA, and other environmental systems where certified reference is not required.