Learn More
Perkin Elmer US LLC Indium - Calibration Standard, 156.60°C
Catalog No. 502113486
Encompass_Preferred
PerkinElmer reference standards are 99.999% pure. DSC instruments require elements of this purity to ensure accurate calibration. Depend on PerkinElmer for quality calibration reference standards.
Catalog No. 50-211-3486 Supplier Perkin Elmer US LLC Supplier No. 03190033
PerkinElmer reference standards are 99.999% pure. DSC instruments require elements of this purity to ensure accurate calibration. Depend on PerkinElmer for quality calibration reference standards.