Learn More
Sigma Aldrich Fine Chemicals Biosciences Indium foil, 10mm disks, thickness 0.125mm, as rolled, 100% | Purity: 100% | Mol Wt: 114.82 | 7440-74-6 | MFCD00134048 | 10EA
Catalog No. 501772242
Encompass
Indium foil, 10mm disks, thickness 0.125mm, as rolled, 100% | Purity: 100% | Mol Wt: 114.82 | 7440-74-6 | MFCD00134048 | 10EA
Catalog No. 50-177-2242 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GF7651636410EA
Indium foil, 10mm disks, thickness 0.125mm, as rolled, 100% | Purity: 100% | Mol Wt: 114.82 | 7440-74-6 | MFCD00134048 | 10EA