Learn More
Strem, An Ascensus Company CAS# 7440-74-6. 100 x 100mm. Indium foil (99.99%). MFCD00134048
SDP
Catalog No. 5090112561
Encompass
CAS# 7440-74-6. 100 x 100mm. Indium foil (99.99%). MFCD00134048. Molecular Weight: 114.76. Molecular Formula: In. Color/form: 0.127mm thick (~2.3g/50 x 50mm) Strem# 49-0050. http://www.strem.com/catalog/v/49-0050/
Catalog No. 50-901-12561 Supplier Strem, An Ascensus Company Supplier No. 490050100X100MM
CAS# 7440-74-6. 100 x 100mm. Indium foil (99.99%). MFCD00134048. Molecular Weight: 114.76. Molecular Formula: In. Color/form: 0.127mm thick (~2.3g/50 x 50mm) Strem# 49-0050. http://www.strem.com/catalog/v/49-0050/