Learn More
Sigma Aldrich Fine Chemicals Biosciences Indium foil, not light tested, 25x25mm, thickness 0.03mm, 99.8% | 7440-74-6 | MFCD00134048 | 5EA
Catalog No. 501772249
Encompass
Indium foil, not light tested, 25x25mm, thickness 0.03mm, 99.8% | Purity: 99.85 | Mol Wt: 114.82 | 7440-74-6 | MFCD00134048 | 5EA
Catalog No. 50-177-2249 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. GF785170425EA
Indium foil, not light tested, 25x25mm, thickness 0.03mm, 99.8% | Purity: 99.85 | Mol Wt: 114.82 | 7440-74-6 | MFCD00134048 | 5EA