Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium(I) bromide, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog No. AA4286506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4286506 5 g
AA4286514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4286506 Supplier Thermo Scientific Chemicals Supplier No. 04286506
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 14280-53-6 | Molecular Formula: InBr

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14280-53-6
Molecular Formula InBr
MDL Number MFCD00064630
Synonym indium i bromide, indium 1+ ;bromide, acmc-1bzdu, indium bromide inbr 6ci,7ci,8ci,9ci

Specifications

Boiling Point 662°C
Melting Point 220°C
Physical Form Polycrystalline Lump
Quantity 5 g
Odor Odorless
Sensitivity Moisture sensitive
Density 4.96 g/mL
UN Number UN3260
Formula Weight 194.73
Percent Purity 99.999%
Grade Puratronic
Assay (metals basis)
Chemical Name or Material Indium (I) bromide
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 238-172-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.