Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium(I) chloride, anhydrous, 99.995% (metals basis)

Catalog No. AA4003314
Change view
Click to view available options
Quantity:
0.5 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4003314 25 g
AA4003302 0.5 g
AA4003306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4003314 Supplier Thermo Scientific Chemicals Supplier No. 04003314
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 13465-10-6 | ClIn | 150.27 g/mol

Indium chloride is a lewis acid catalyst in organic reactions such as Friedel-Crafts acylations and Diels-Alder reactions and it is used in high-performing solar cells such as optics and microelectronics. It applications finds in semiconductor technology, producing phosphor,low pressure sodium lamp and produce high purity chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13465-10-6
Molecular Formula ClIn
Molecular Weight (g/mol) 150.27
MDL Number MFCD00148875
InChI Key APHGZSBLRQFRCA-UHFFFAOYSA-M
IUPAC Name λ¹-indiganyliumylidene chloride
SMILES [Cl-].[In+]

Specifications

Boiling Point 608°C
Melting Point ∼225°C
Physical Form Crystalline
Quantity 25 g
UN Number UN3260
Beilstein EM>13,
Solubility Information Soluble in water and THF.
Formula Weight 150.27
Percent Purity 99.995%
Odor Chlorine-like
Sensitivity Moisture sensitive
Density 4.19 g/mL
Assay (metals basis)
Chemical Name or Material Indium(I) chloride, Anhydrous
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
EINECSNumber 236-693-3
TSCA Yes
storageNote1
  • Packaged under argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.