Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Indium(I) iodide

Catalog No. 4136585G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
41-365-85G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 41-365-85G Supplier Sigma Aldrich Supplier No. 4136585G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Has been discovered and used as a reagent to propagate some organic reactions, such as cyclization and cleavage of diselenide or disulfide. Can also be used to prepare In-containing complexes.AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 13966-94-4
Quantity 5 g
Molecular Formula IIn
Linear Formula InI
MDL Number MFCD00135543
Synonym Indium monoiodide
Molecular Weight (g/mol) 241.72
Percent Purity 99.999% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.