Learn More
Indium(I) iodide, ultra dry, 99.998% (metals basis)
Description
Indium(I) iodide can be used to prepare indium cluster and chain compounds. It is also used as a reagent to propagate organic reactions viz. cyclization and cleavage of diselenide or disulfide. It plays an important role in preparing indium containing complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Ampouled under argon
Specifications
Specifications
| Melting Point | 351°C |
| Physical Form | Beads |
| Quantity | 1 g |
| Packaging | Ampule |
| Sensitivity | Light and moisture sensitive |
| Solubility Information | Slightly soluble in some organic solvents,pyridine or m-xylene mixture. |
| Formula Weight | 241.72 |
| Percent Purity | 99.998% |
| Assay | (metals basis) |
| Chemical Name or Material | Indium(I) iodide, Ultra dry |
Safety and Handling
| Hazard Category | H302-H315-H317-H319-H334-H335 |
| Hazard Statement | GHS H Statement H334-H302-H335-H315-H319-H317 May cause allergy or asthma symptoms or breathing difficulties if inhaled. Harmful if swallowed. May cause respiratory irritation. Causes skin irritation. Causes serious eye irritation. May cause an allergic skin reaction. |
| Precautionary Statement | P261-P264b-P270-P271-P272-P280-P285-P301+P312-P302+P352-P304+P340-P305+P351+P338-P330-P333+P313-P342+P311-P362-P501c |
| EINECSNumber | 237-746-3 |
| TSCA | Yes |
| storageNote1 |
|
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.