Learn More
Crescent Chemical Co Inc INDIUM ICP STD 100ML
SDP
Catalog No. 5072204
Encompass_Preferred
Indium, HNO3 Single Element Plasma Emission Spectroscopy Standards 10,000 UgmL NIST Traceable CAS Size - 100mL Catalog Number - 8695
Catalog No. 50-722-04 Supplier Crescent Chemical Co Inc Supplier No. IS8695 (100ML)
Add to Cart
Add to Cart
Indium, HNO3 Single Element Plasma Emission Spectroscopy Standards 10,000 UgmL NIST Traceable CAS Size - 100mL Catalog Number - 8695