Learn More
Strem, An Ascensus Company CAS# 25114-58-3. 5g. Indium(III) acetate (99.99%-In) PURATREM. MFCD00054347
SDP
Supplier:  Strem, An Ascensus Company 9349365G
Encompass
CAS# 25114-58-3. 5g. Indium(III) acetate (99.99%-In) PURATREM. MFCD00054347. Molecular Weight: 291.95. Molecular Formula: In(OOCCH3)3. Color/form: white xtl. Strem# 93-4936. http://www.strem.com/catalog/v/93-4936/
Catalog No. 50-901-15527