Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium(III) bromide, anhydrous, 99.99% (metals basis)

Catalog No. AA4012609
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4012609 10 g
AA4012618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4012609 Supplier Thermo Scientific Chemicals Supplier No. 04012609
Add to Cart

CAS: 13465-09-3

Indium(III) bromide is used in organic synthesis as a water tolerant Lewis acid.It efficiently catalyzes the three-component coupling of β-keto esters, aldehydes and urea (or thiourea) to afford the corresponding dihydropyrimidinones. It is used as a catalyst to produce dithioacetals when unactivated alkynes react with thiols and fields such as optics and microelectronics that utilize semiconductor technology have wide uses for indium in high-performing solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13465-09-3
MDL Number MFCD00011057

Specifications

Melting Point ∼436°C
Color White to Yellow
Physical Form Crystalline Powder and/or Chunks
Quantity 10 g
Linear Formula InBr3
Solubility Information Soluble in water.
Formula Weight 354.55
Percent Purity 99.99%
Odor Odorless
Sensitivity Hygroscopic
Density 4.74 g/mL
Assay (metals basis)
Chemical Name or Material Indium(III) bromide, Anhydrous
Show More Show Less
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-692-8
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.