Learn More
Indium(III) bromide, anhydrous, 99.99% (metals basis)
Description
Indium(III) bromide is used in organic synthesis as a water tolerant Lewis acid.It efficiently catalyzes the three-component coupling of β-keto esters, aldehydes and urea (or thiourea) to afford the corresponding dihydropyrimidinones. It is used as a catalyst to produce dithioacetals when unactivated alkynes react with thiols and fields such as optics and microelectronics that utilize semiconductor technology have wide uses for indium in high-performing solar cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | ∼436°C |
| Color | White to Yellow |
| Physical Form | Crystalline Powder and/or Chunks |
| Quantity | 50 g |
| Odor | Odorless |
| Sensitivity | Hygroscopic |
| Density | 4.74 g/mL |
| Linear Formula | InBr3 |
| Solubility Information | Soluble in water. |
| Formula Weight | 354.55 |
| Show More |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 236-692-8 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.