Learn More
Strem, An Ascensus Company CAS# 10025-82-8. 5g. Indium(III) chloride, anhydrous (99.999%-In) PURATREM. MFCD00011058
SDP
Supplier:  Strem, An Ascensus Company 9349325G
Encompass
CAS# 10025-82-8. 5g. Indium(III) chloride, anhydrous (99.999%-In) PURATREM. MFCD00011058. Molecular Weight: 221.18. Molecular Formula: InCl3. Color/form: white pwdr. Strem# 93-4932. http://www.strem.com/catalog/v/93-4932/
Catalog No. 50-901-15524