Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium(III) fluoride, anhydrous, 99.95% (metals basis)

Catalog No. AA4552514
Click to view available options
Quantity:
5 g
25 g

CAS: 7783-52-0 | F3In

Indium(III) fluoride is used in the preparation of non-oxide glasses. It acts as a catalyst in the reaction of trimethylsilyl cyanide (TMSCN) to aldehydes to get cyanohydrins. It play a vital role as a catalyst in organic synthesis to prepare fluorine containing 2,4,5-trisubstituted imidazoles and polysubstituted pyrroles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-52-0
Molecular Formula F3In
MDL Number MFCD00016149
Synonym indium fluoride, indium trifluoride, indium 3+ trifluoride, acmc-1bcyn, indium 3+ ion trifluoride, fluoride, fluoride, in, fluoride

Specifications

Boiling Point >1200°C
Melting Point ∼1170°C
Color White
Physical Form Powder
Quantity 25 g
Linear Formula InF3
Merck Index 14,4959
Solubility Information Slightly soluble in water. Freely soluble in dilute acids.
Formula Weight 171.82
Percent Purity 99.95%
Odor Odorless
Sensitivity Moisture sensitive
Density 4.39 g/mL
Assay (metals basis)
Chemical Name or Material Indium (III) fluoride, Anhydrous
Show More Show Less
Hazard Category H302+H332-H314
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 232-005-0
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.