Learn More
Medchemexpress LLC HY-133023 10mg Medchemexpress, Indium(III) isopropoxide CAS:38218-24-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13302310MG
Encompass_Preferred
Medchemexpress, HY-133023 10mg Indium(III) isopropoxide CAS:38218-24-5 Indium(III) Isopropoxide is an organo-metallic compound. Indium(III) Isopropoxide uesd as a hydrogen transfer catalyst for conversion of benzylic alcohols into aldehydes or ketones via Oppenauer oxidation. Indium(III) Isopropoxide also can be used as metal precursor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9844