Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Indium(III) nitrate hydrate

Catalog No. 32612750G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
32-612-750G 50 g
32-612-710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 32-612-750G Supplier Sigma Aldrich Supplier No. 32612750G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Indium(III) nitrate hydrate is an indium based precursor solution, which can be annealed at 500°C and spin coated to form indium oxide thin films.Indium(III) nitrate hydrate is used in the preparation of indium based substrates for the fabrication of semiconductor devices, such as thin film transistors.

Specifications

Quantity 50 g
Molecular Formula InN3O9 -+ xH2O
Linear Formula In(NO3)3 -+ xH2O
MDL Number MFCD00149742
Molecular Weight (g/mol) 300.83
Percent Purity 99.99% trace metals basis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.