Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium(III) nitrate hydrate, 99.99% (metals basis)

Catalog No. AA4018518
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4018518 50 g
AA4018509 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4018518 Supplier Thermo Scientific Chemicals Supplier No. 04018518
Only null left
Add to Cart

CAS: 207398-97-8 | InN3O9 | 300.83 g/mol

Indium(III) nitrate hydrate acts as an oxidizing agent. It is used in the manufacture of coordinated polymer films which are used in the preparation of indium(III) oxide nanoparticles. It is used in the synthesis of other indium complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207398-97-8
Molecular Formula InN3O9
Molecular Weight (g/mol) 300.83
MDL Number MFCD00149742
InChI Key LKRFCKCBYVZXTC-UHFFFAOYSA-N
Synonym indium iii nitrate hydrate, indium nitrate hydrate, nitric acid, indium 3+ salt, hydrate 2:9, indium 3+ ion hydrate trinitrate, indium nitrate hydrate 4.5, indiumnitratehexahydrate, indium nitrate, hydrous, in no3 33h2o, indiumnitratehydratewhitextl, in.3no3.h2o
PubChem CID 3084148
SMILES [In+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 100°C-H2O
Color White
Physical Form Lump/Powder
Quantity 50 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Soluble in water,methanol and ethanol.
IUPAC Name indium(3+) trinitrate
Formula Weight 300.83 (Anhydrous)
Sensitivity Hygroscopic
Appearance White to off-white lump/powder
Chemical Name or Material Indium(III) nitrate hydrate
Show More Show Less
Hazard Category H272-H315-H319
Precautionary Statement P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
DOTInformation Hazard Class: 5.1; Packaging Group: III
EINECSNumber 237-393-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.