Learn More
Indium(III) nitrate hydrate, Puratronic™, 99.999% (metals basis)
Description
Indium(III) nitrate hydrate acts as an oxidizing agent. It is used in the manufacture of coordinated polymer films which are used in the preparation of indium(III) oxide nanoparticles. It is used in the synthesis of other indium complexes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | 100°C (in -H2O) |
Color | White |
Physical Form | Crystalline Aggregates |
Quantity | 100 g |
UN Number | UN1477 |
Solubility Information | Soluble in water,methanol and ethanol. |
IUPAC Name | indium(3+);trinitrate;hydrate |
Formula Weight | 300.83 (Anhydrous) |
Percent Purity | 99.999% |
Grade | Puratronic™ |
Show More |
Safety and Handling
Hazard Category | H272-H315-H319 |
Hazard Statement | GHS H Statement H272-H315-H319 May intensify fire oxidizer. Causes skin irritation. Causes serious eye irritation. |
Precautionary Statement | P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c |
DOTInformation | Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: NITRATES, INORGANIC, N.O.S. |
EINECSNumber | 237-393-5 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.