Indium(III) oxide, 99.9% (metals basis), Alfa Aesar™

Manufacturer: Alfa Aesar 1258130

Quantity 250g

 View more versions of this product

Catalog No. AA1258130

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1312-43-2
    Physical Form Powder
    Assay Percent Range 99.994% (Metal basis)
    Molecular Formula In2O3
    Formula Weight 277.64
    MDL Number MFCD00011060
    Chemical Name or Material Indium(III) oxide

    Specifications

    Formula Weight 277.64
    Quantity 250g
    Color Yellow
    EINECS Number 215-193-9
    Physical Form Powder
    Odor Odorless
    Assay Percent Range 99.994% (Metal basis)
    Molecular Formula In2O3
    Appearance Yellow powder
    MDL Number MFCD00011060
    CAS 1312-43-2
    Chemical Name or Material Indium(III) oxide

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit