Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium(III) oxide, 99.994% (metals basis)

Catalog No. AA1185330
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1185330 250 g
AA1185309 10 g
AA1185318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1185330 Supplier Thermo Scientific Chemicals Supplier No. 01185330
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 1312-43-2 | In2O3 | 277.63 g/mol

Indium(III) oxide is used as a chemical reagent and intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1312-43-2
Molecular Formula In2O3
Molecular Weight (g/mol) 277.63
MDL Number MFCD00011060
InChI Key PJXISJQVUVHSOJ-UHFFFAOYSA-N
IUPAC Name diindium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[In+3].[In+3]

Specifications

Melting Point 850° volat.
Color Yellow
Physical Form Powder
Quantity 250 g
Odor Odorless
Assay Percent Range 99.994% (Metals basis)
Density 7.18
Solubility Information Insoluble in water.
Formula Weight 277.64
Chemical Name or Material Indium(III) oxide
EINECSNumber 215-193-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.