Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Indium(III) oxide, 99.999% (metals basis)
Click to view available options
Quantity:
10g
50g
Specifications
Merck Index | 14,4952 |
Solubility Information | Insoluble in water. |
Formula Weight | 277.64 |
Physical Form | 3-6mm Sintered Granules |
Percent Purity | 99.999% |
Density | 7.18 g/mL |
Odor | Odorless |
Particle Size | 3 to 6mm |
Melting Point | 850°C (volat.) |
Quantity | 10g |
Show More |
Safety and Handling
EINECSNumber : 215-193-9
TSCA : Yes
Recommended Storage : Ambient temperatures