Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium(III) phosphate, 98%

Catalog No. AAA1828106
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1828106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1828106 Supplier Thermo Scientific Chemicals Supplier No. A1828106
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 14693-82-4 | InO4P | 209.788 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14693-82-4
Molecular Formula InO4P
Molecular Weight (g/mol) 209.788
MDL Number MFCD00016154
InChI Key UJXZVRRCKFUQKG-UHFFFAOYSA-K
Synonym indium iii phosphate, acmc-1cjl6, indium 3+ phosphate, indium 3+ ion phosphate
PubChem CID 9815692
IUPAC Name indium(3+);phosphate
SMILES [O-]P(=O)([O-])[O-].[In+3]

Specifications

Quantity 5 g
Formula Weight 209.79
Percent Purity 98%
Density 4.9
Chemical Name or Material Indium(III) phosphate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.