Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Indium(III) selenide

Catalog No. 4033185G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-331-85G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 40-331-85G Supplier Sigma Aldrich Supplier No. 4033185G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Indium selenide (In2Se3) exists in 4 crystal structures .The metallic α-phase of indium selenide converts to intrinsic conduction α-phase under change in temperature. In2Se3 can be prepared by stoichiometric combination of the elements at 1000 to 1100oC temperatures. Influence of phase transformations on its optical and electrical properties has been reported.

Specifications

CAS 12056-07-4
Quantity 5 g
Molecular Formula In2Se3
Linear Formula In2Se3
MDL Number MFCD00016147
Synonym Indium selenide; Indium sesquiselenide
Molecular Weight (g/mol) 466.52
Density 5.67 g/mL (at 25°C)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.