Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium(III) sulfide, 99.999% (metals basis)

Catalog No. AA4556309
Change view
Click to view available options
Quantity:
2 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4556309 10 g
AA4556304 2 g
AA4556318 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4556309 Supplier Thermo Scientific Chemicals Supplier No. 04556309
Only null left
Add to Cart

CAS: 12030-24-9 | In2S3 | 325.82 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12030-24-9
Molecular Formula In2S3
Molecular Weight (g/mol) 325.82
MDL Number MFCD00011061
InChI Key SIXIBASSFIFHDK-UHFFFAOYSA-N
IUPAC Name diindium(3+) trisulfanediide
SMILES [S--].[S--].[S--].[In+3].[In+3]

Specifications

Melting Point 1050°C
Physical Form Powder
Quantity 10 g
Formula Weight 325.83
Percent Purity 99.999%
Odor Like rotten eggs
Sensitivity Moisture sensitive
Assay (metals basis)
Chemical Name or Material Indium (III) sulfide
Hazard Category H302+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H332-H315-H319-H335
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 234-742-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.