Learn More
Strem, An Ascensus Company CAS# 128008-30-0. 5g. Indium(III) trifluoromethanesulfonate, 99% (Indium triflate). MFCD00144478
SDP
Catalog No. 5090112567
Encompass
CAS# 128008-30-0. 5g. Indium(III) trifluoromethanesulfonate, 99% (Indium triflate). MFCD00144478. Molecular Weight: 562.02. Molecular Formula: In(CF3SO3)3. Color/form: white pwdr. Strem# 49-1800. http://www.strem.com/catalog/v/49-1800/
Catalog No. 50-901-12567 Supplier Strem, An Ascensus Company Supplier No. 4918005G
CAS# 128008-30-0. 5g. Indium(III) trifluoromethanesulfonate, 99% (Indium triflate). MFCD00144478. Molecular Weight: 562.02. Molecular Formula: In(CF3SO3)3. Color/form: white pwdr. Strem# 49-1800. http://www.strem.com/catalog/v/49-1800/