Learn More
Sigma Aldrich Indium Phosphide Zinc Sulfide (inp/zns) Quantum Dots In Toluene Stabilized With Oleic Acid Fluorescence ?em 530 Nm
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
Specifications
Specifications
| CAS | 2398-8-0--7 |
| Linear Formula | InP/ZnS |
| Quantity | 10 mg |
| Synonym | Fluorescent nanocrystals; QDs |
| Biological Activity | Central nervous system, Lungs |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.