Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Indium Phosphide Zinc Sulfide (inp/zns) Quantum Dots In Toluene Stabilized With Oleic Acid Fluorescence ?em 620 Nm

Catalog No. 92029010MG Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
92-029-010MG 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 92-029-010MG Supplier Sigma Aldrich Supplier No. 92029010MG
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Reactive group: AmineInP/ZnS core shell nanocrystals are Cadmium free/heavy metal free quantum dots suitable for different applications like light emitting diodes such as LEDs, displays, and biomedical applications. These Qdots are environmentally-friendly, heavy metal-free alternative to cadmium based quantum dots. These InP/ZnS core/shell quantum dots are non-toxic while offering high performance for various applications

Specifications

CAS 2398-8-0--7
Linear Formula InP/ZnS
Quantity 10 mg
Synonym Fluorescent nanocrystals; QDs
Biological Activity Central nervous system, Lungs
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.