Learn More
Strem, An Ascensus Company CAS# 7440-74-6. 5g. Indium powder (99.99%). MFCD00134048
SDP
Catalog No. 5090115529
Encompass
CAS# 7440-74-6. 5g. Indium powder (99.99%). MFCD00134048. Molecular Weight: 114.76. Molecular Formula: In. Color/form: -325 mesh Strem# 93-4940. http://www.strem.com/catalog/v/93-4940/
Catalog No. 50-901-15529 Supplier Strem, An Ascensus Company Supplier No. 9349405G
CAS# 7440-74-6. 5g. Indium powder (99.99%). MFCD00134048. Molecular Weight: 114.76. Molecular Formula: In. Color/form: -325 mesh Strem# 93-4940. http://www.strem.com/catalog/v/93-4940/