Learn More
Strem, An Ascensus Company CAS# 7440-74-6. 25g. Indium shot (99.9999%). MFCD00134048
SDP
Catalog No. 5090115536
Encompass
CAS# 7440-74-6. 25g. Indium shot (99.9999%). MFCD00134048. Molecular Weight: 114.76. Molecular Formula: In. Color/form: 4-8 mm Strem# 93-4943. http://www.strem.com/catalog/v/93-4943/
Catalog No. 50-901-15536 Supplier Strem, An Ascensus Company Supplier No. 93494325G
CAS# 7440-74-6. 25g. Indium shot (99.9999%). MFCD00134048. Molecular Weight: 114.76. Molecular Formula: In. Color/form: 4-8 mm Strem# 93-4943. http://www.strem.com/catalog/v/93-4943/