Learn More
SPEX CERTIPREP LLC 3P INDIUM STD 1000UG/ML 100ML
Catalog No. NC0283453
Encompass_Preferred
(HPS by Zeptometrix) In 1000ug/mL in 2% HNO3 Exp Date: 18 Months This standard is traceable to NIST and certified Guide 34 and 17025.
Catalog No. NC0283453 Supplier SPEX CERTIPREP LLC 3P Supplier No. 1000241100
(HPS by Zeptometrix) In 1000ug/mL in 2% HNO3 Exp Date: 18 Months This standard is traceable to NIST and certified Guide 34 and 17025.