Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Indium tin oxide coated glass slide, rectangular

Catalog No. 57635225PAK Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
10 pak
25 pak
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-635-225PAK 25 pak
57-635-210PAK 10 pak
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 57-635-225PAK Supplier Sigma Aldrich Supplier No. 57635225PAK
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Indium tin oxide (ITO) is widely used as transparent conducting oxides. Its major characteristics are electrical conductivity, optical transparency and the ease to be deposited as films.ITO cover slides may be used for closed digital microfluidic devices and as a transparent electrodes in display and other optoelectronic devices. Some other potential uses that have been reported are:Eumelanin samples prepared on ITO covered glass substrates. Interface windows of the liquid cell for optical beam deflection (OBD) sensing contain a 20-nm-thick indium tin oxide film.Mesoporous TiO2 films were formed on ITO conductive glass substrates.

Specifications

CAS 50926-11-9
Quantity 25 pak
Linear Formula In2O3 -+ (SnO2)x
MDL Number MFCD00171662
Synonym ITO; ITO coated slide, rectangular
Refractive Index n20/D 1.517 (literature)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.