Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Indium tin oxide coated glass slide, rectangular

Catalog No. 57827425PAK Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 pak
25 pak
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
57-827-425PAK 25 pak
57-827-410PAK 10 pak
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 57-827-425PAK Supplier Sigma Aldrich Supplier No. 57827425PAK
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Indium tin oxide (ITO) is widely used as transparent conducting oxides. Its major characteristics are electrical conductivity, optical transparency and the ease to be deposited as films.ITO was used to mount frozen sections of mouse brain and heart.ITO cover slides may be used for closed digital microfluidic devices. Some other uses that have been reported are:Eumelanin samples prepared on ITO covered glass substrates. Interface windows of the liquid cell for optical beam delection (OBD) sensing contain a 20-nm-thick indium tin oxide filmMesoporous TiO2 films were formed on ITO conductive glassesI

Specifications

CAS 50926-11-9
Quantity 25 pak
Linear Formula In2O3 -+ (SnO2)x
MDL Number MFCD00171662
Synonym ITO; ITO coated slide, rectangular
Refractive Index n20/D 1.517 (literature)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.