Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Indium tin oxide coated glass slide, square

Catalog No. 70319210PAK Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 pak
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-319-210PAK 10 pak
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 70-319-210PAK Supplier Sigma Aldrich Supplier No. 70319210PAK
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

substrate: unpolished float glass, SiO2 passivated (200-300 +a)ITO glass slides were used as a substrate for fabricating polymer solar cells based on blends of polymeric semiconductors and organic light emitting diodes (OLEDS). ITO substrates can also be used to design transparent heater arrays by etching the surface of the substrate by a printing tool. ITO cover slides may be used for closed digital microfluidic devices and as transparent electrodes in display and other optoelectronic devices.Some other probable uses that has been reported are:Eumelanin samples prepared on ITO covered glass substrates. Interface windows of the liquid cell for optical beam deflection (OBD) sensing contains a 20-nm-thick indium tin oxide film.Mesoporous TiO2 films were formed on ITO conductive glasses.

Specifications

CAS 50926-11-9
Quantity 10 pak
Linear Formula In2O3 -+ (SnO2)x
MDL Number MFCD00171662
Synonym ITO; ITO coated slide, square
Refractive Index n20/D 1.517
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.