Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Indium tin oxide coated PET

Catalog No. 6393035EA Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 Ea.
5 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
63-930-35EA 5 Ea.
63-930-31EA 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 63-930-35EA Supplier Sigma Aldrich Supplier No. 6393035EA
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Indium tin oxide films are inherently brittle in nature. To overcome this, ITO is coated with transparent conductive polymers or nanomaterial. Indium tin oxide coated polyethylene terephthalate (PET) film is optically transparent and electrically conductive.The ITO coating is protected by a thin, transparent film, that can be easily peeled off.A radius of curvature of up to 75 mm will not damage the ITO coating.Micro mechanical properties of ITO-PET were investigated. Electrical resistance of ITO-PET increases in case of degradation. ITO-PET was used as an electrode in the electrochemical measurements in a study of biosensors composed of polymer brushes. ITO microelectrodes were studied as an electronic tongue for analyte recognition. The sheet resistance of ITO, after bending to a radius of 25 mm was investigated, for its use for flexible optical devices. Giant unilamellar vesicle (GUVs) were produced on ITO coated glass slides by drying droplets of the FhuA proteoliposomes.

Specifications

Quantity 5 Ea.
Linear Formula In2O3 -+ (SnO2)x
MDL Number MFCD00171662
Synonym ITO-PET; Polyethylene terephthalate film, ITO coated
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.