Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Indium tin oxide, Vacuum Deposition Grade, 99.99% (metals basis)

Catalog No. AA4267706
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4267706 5 g
AA4267714 25 g
AA4267722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA4267706 Supplier Thermo Scientific Chemicals Supplier No. 04267706
Add to Cart

CAS: 50926-11-9 | InOSn | 249.527 g/mol

Indium tin oxide is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50926-11-9
Molecular Formula InOSn
Molecular Weight (g/mol) 249.527
MDL Number MFCD00171662
InChI Key AMGQUBHHOARCQH-UHFFFAOYSA-N
Synonym acmc-1au9m, tin ii oxide indigane
PubChem CID 57350325
IUPAC Name indium;oxotin
SMILES O=[Sn].[In]

Specifications

Physical Form Pieces
Quantity 5 g
Solubility Information Insoluble in water.
Percent Purity 99.99%
Assay (metals basis)
Chemical Name or Material Indium tin oxide
Hazard Category H335-H351
Hazard Statement GHS H Statement
H335
May cause respiratory irritation.
Precautionary Statement P201-P202-P261-P271-P281-P304+P340-P308+P313-P501c
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.