Learn More
Sigma Aldrich Fine Chemicals Biosciences IndiumIII acetate 99.99 tr10G
Catalog No. 502899282
Encompass
Indium(III) acetate is water soluble and decomposes to indium oxide on heating.
Catalog No. 50-289-9282 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 51027010G
Indium(III) acetate is water soluble and decomposes to indium oxide on heating.