Learn More
Sigma Aldrich Fine Chemicals Biosciences IndiumIII acetate 99.99 tr50G
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 51027050G
Encompass
Indium(III) acetate is water soluble and decomposes to indium oxide on heating.
Catalog No. 50-290-3947