Learn More
Description
- Used in extremely stable environments
- 1mg to 500mg weights are one-piece construction with no adjusting cavity with one end turned up for easy handling
- 1g to 5kg are two-piece construction with handling knob
- Surfaces are polished to a perfect, mirror like finish
- Can be used as a reference standard in calibrating other weights
- Appropriate for calibrating high-precision analytical balances with a readability as low as 0.1 mg to 0.01mg
Certifications
A NIST/NVLAP™ weight calibration certificate or traceable certificate may be purchased to meet all of your ISO™, GLP, GMP, FDA, or nuclear requirements
Specifications
Specifications
| Product Type | One Piece Metric Analytical Sheet Weight |
| Classification | ASTM Class 1 |
| Material | Stainless Steel |
| Weight (Metric) | 500 mg |
| Density | 7.95g/cm3 at 20°C |
| Mass | 500 mg |
| Shape | Flat with one end turned up for easy handling |
| Includes | Polycarbonate case |
| Quantity | Each |
| Packaging Quantity | 1 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.